siRNA Details
| Gene Symbol: Vmn1r236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134201.2 | NP_598962.2 | 1 | 534 | ||
| GenBank mRNA | BC125459.1 | - | 1 | 534 | 40129974 | |
| BC125461.1 | - | 1 | 534 | 40129976 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r236| Gene Symbol: Vmn1r236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134201.2 | NP_598962.2 | 1 | 534 | ||
| GenBank mRNA | BC125459.1 | - | 1 | 534 | 40129974 | |
| BC125461.1 | - | 1 | 534 | 40129976 | ||