siRNA Details
| Gene Symbol: Vmn1r67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134229.2 | NP_598990.2 | 1 | 381 | ||
| GenBank mRNA | BC138174.1 | - | 1 | 381 | 8861193 | |
| BC145742.1 | - | 1 | 381 | 40131045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r67| Gene Symbol: Vmn1r67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134229.2 | NP_598990.2 | 1 | 381 | ||
| GenBank mRNA | BC138174.1 | - | 1 | 381 | 8861193 | |
| BC145742.1 | - | 1 | 381 | 40131045 | ||