siRNA Details
| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102468.1 | NP_001095938.1 | 3 | 207 | ||
| NM_138304.2 | NP_612177.1 | 4 | 348 | |||
| GenBank mRNA | AK008039.1 | - | 4 | 334 | ||
| AK146656.1 | - | 4 | 321 | |||
| AY061807.1 | - | 4 | 348 | |||
| BC013541.1 | - | 4 | 326 | 4241711 | ||
| BC023475.1 | - | 4 | 313 | 5042406 | ||
| BY097052.1 | - | 3 | 220 | |||

Calml4
Genomic Map