siRNA Details
| Gene Symbol: Spg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138584.2 | NP_613050.1 | 8 | 955 | ||
| GenBank mRNA | AK009332.1 | - | 8 | 947 | ||
| AK011985.1 | - | 8 | 922 | |||
| AK090017.1 | - | 8 | 943 | |||
| AK149930.1 | - | 8 | 929 | |||
| AK150215.1 | - | 8 | 956 | |||
| AK150581.1 | - | 8 | 933 | |||
| AK153284.1 | - | 8 | 917 | |||
| AK170666.1 | - | 8 | 948 | |||
| AK197543.1 | - | 2 | 71 | |||
| AK212652.1 | - | 2 | 71 | |||
| AK216945.1 | - | 2 | 71 | |||
| BC099434.1 | - | 7 | 720 | 30714184 | ||
| BC132377.1 | - | 8 | 932 | 40130654 | ||
| BC132403.1 | - | 8 | 932 | 40130680 | ||

Spg21
Genomic Map