siRNA Details
| Gene Symbol: Exosc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138586.3 | NP_613052.1 | 2 | 286 | ||
| GenBank mRNA | AK006475.1 | - | 2 | 239 | ||
| AK007372.1 | - | 2 | 275 | |||
| AK137780.1 | - | 249 | ||||
| AK155228.1 | - | 2 | 238 | |||
| BB856072.1 | - | 2 | 288 | |||
| BC034358.1 | - | 2 | 232 | 2651418 | ||
| BC092393.1 | - | 2 | 227 | |||
| BY196593.1 | - | 2 | 284 | |||
| BY198385.1 | - | 2 | 284 | |||

Exosc5
Genomic Map