siRNA Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127338.1 | NP_001120810.1 | 5 | 534 | ||
| NM_138600.4 | NP_613066.2 | 3 | 340 | |||
| GenBank mRNA | AK004991.1 | - | 5 | 468 | ||
| AK145873.1 | - | 3 | 283 | |||
| AK160117.1 | - | 3 | 267 | |||
| AK169195.1 | - | 3 | 283 | |||
| AK187582.1 | - | 2 | 83 | |||
| BC012407.1 | - | 3 | 302 | 4191228 | ||
| BY017676.1 | - | 3 | 340 | |||

Aldh7a1
Genomic Map