siRNA Details
| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138628.3 | NP_619534.2 | 10 | 2639 | ||
| GenBank mRNA | AF422245.1 | - | 10 | 2576 | ||
| AK137442.1 | - | 10 | 2554 | |||
| BC141023.1 | - | 10 | 2552 | 9055635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txlnb| Gene Symbol: Txlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138628.3 | NP_619534.2 | 10 | 2639 | ||
| GenBank mRNA | AF422245.1 | - | 10 | 2576 | ||
| AK137442.1 | - | 10 | 2554 | |||
| BC141023.1 | - | 10 | 2552 | 9055635 | ||