siRNA Details
| Gene Symbol: Nap1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138742.1 | NP_620081.1 | 1 | 734 | ||
| GenBank mRNA | AB010711.1 | - | 1 | 750 | ||
| AK078521.1 | - | 1 | 725 | |||
| BC020176.1 | - | 1 | 734 | 4505496 | ||
| BC027045.1 | - | 1 | 631 | 5363560 | ||

Nap1l3
Genomic Map