siRNA Details
Gene Symbol: Spag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139151.4 | NP_631890.3 | 2 | 496 | ||
GenBank mRNA | AB041554.1 | - | 2 | 576 | ||
AK028080.1 | - | 2 | 465 | |||
AK043812.1 | - | 2 | 464 | |||
AK220494.1 | - | 973 | ||||
BC107321.1 | - | 2 | 395 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
