siRNA Details
| Gene Symbol: Asb18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139152.1 | NP_631891.1 | 1 | 95 | ||
| GenBank mRNA | AY057054.1 | - | 1 | 95 | ||
| BC107370.1 | - | 1 | 95 | |||
| BC107371.1 | - | 1 | 95 | |||
| BC127152.1 | - | 1 | 95 | |||
| BC141295.1 | - | 1 | 184 | 9055907 | ||
| BC145621.1 | - | 1 | 184 | 9054189 | ||

Asb18
Genomic Map