siRNA Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139154.2 | NP_631893.1 | 4 | 333 | ||
| GenBank mRNA | AB232639.1 | - | 3 | 227 | ||
| AF422144.1 | - | 3 | 286 | |||
| AK133766.1 | - | 3 | 109 | |||
| BC027008.1 | - | 4 | 321 | 5364768 | ||
| BC037461.1 | - | 4 | 454 | |||

Rab40c
Genomic Map