siRNA Details
| Gene Symbol: Dppa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139218.1 | NP_631964.1 | 2 | 224 | ||
| GenBank mRNA | AB072734.1 | - | 2 | 125 | ||
| AF490347.1 | - | 2 | 229 | |||
| AK135769.1 | - | 2 | 229 | |||
| AK136263.1 | - | 2 | 229 | |||
| AY082485.1 | - | 2 | 224 | |||
| AY134859.1 | - | 2 | 236 | |||
| BC099433.1 | - | 2 | 202 | 30933569 | ||
| BC100331.1 | - | 2 | 199 | 30934920 | ||
| BC107340.1 | - | 2 | 156 | 40055218 | ||
| BC107341.1 | - | 2 | 156 | 40055220 | ||

Dppa3
Genomic Map