siRNA Details
| Gene Symbol: DCX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000555.3 | NP_000546.2 | 7 | 8262 | ||
| NM_001195553.1 | NP_001182482.1 | 7 | 7982 | |||
| NM_178151.2 | NP_835364.1 | 7 | 8091 | |||
| NM_178152.2 | NP_835365.1 | 7 | 7979 | |||
| NM_178153.2 | NP_835366.1 | 7 | 7964 | |||
| XM_011530878.2 | XP_011529180.1 | 7 | 7939 | |||
| XM_011530879.2 | XP_011529181.1 | 6 | 7856 | |||
| XM_011530880.2 | XP_011529182.1 | 6 | 7841 | |||
| XM_017029312.1 | XP_016884801.1 | 7 | 8047 | |||
| GenBank mRNA | AJ003112.1 | - | 7 | 8277 | ||
| AK002120.1 | - | 1 | 1322 | |||

DCX
Genomic Map