siRNA Details
| Gene Symbol: Phf21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109690.1 | NP_001103160.1 | 5, 6 | 647 | ||
| NM_001109691.1 | NP_001103161.1 | 8, 9 | 803 | |||
| NM_138755.2 | NP_620094.2 | 7, 8 | 806 | |||
| GenBank mRNA | AB105178.1 | - | 7, 8 | 712 | ||
| AK037174.1 | - | 8, 9 | 804 | |||
| AK129424.1 | - | 7, 8 | 2065 | |||
| AK137085.1 | - | 4, 5 | 614 | |||
| AY206982.1 | - | 2, 3 | 291 | |||
| BC019181.1 | - | 7, 8 | 661 | 5008954 | ||

Phf21a
Genomic Map