siRNA Details
| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138757.2 | NP_620096.2 | 3, 4 | 851 | ||
| GenBank mRNA | BC026851.1 | - | 3, 4 | 780 | 4459471 | |
| BC027229.1 | - | 3, 4 | 436 | 2615769 | ||
| CD560819.1 | - | 1, 2 | 58 | |||
| CF167981.1 | - | 3, 4 | 447 | |||

Asun
Genomic Map