siRNA Details
| Gene Symbol: Spats2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139140.1 | NP_631879.1 | 3 | 402 | ||
| GenBank mRNA | AB079132.1 | - | 3 | 402 | ||
| AK048720.1 | - | 3 | 444 | |||
| AK135838.1 | - | 3 | 460 | |||
| AK159335.1 | - | 3 | 496 | |||
| AK167788.1 | - | 2 | 79 | |||
| AK171266.1 | - | 2 | 157 | |||
| BC023432.1 | - | 1 | 52 | 5029821 | ||
| BC072579.1 | - | 3 | 669 | |||

Spats2
Genomic Map