siRNA Details
| Gene Symbol: Slc6a20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ428067.1 | - | 2004 | |||
| AK043297.1 | - | 11 | 2002 | |||
| AK080979.1 | - | 2004 | ||||
| AK140445.1 | - | 2002 | ||||
| AK160468.1 | - | 2002 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a20a| Gene Symbol: Slc6a20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ428067.1 | - | 2004 | |||
| AK043297.1 | - | 11 | 2002 | |||
| AK080979.1 | - | 2004 | ||||
| AK140445.1 | - | 2002 | ||||
| AK160468.1 | - | 2002 | ||||