siRNA Details
| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139147.3 | NP_631886.2 | 6 | 872 | ||
| GenBank mRNA | AB232638.1 | - | 6 | 760 | ||
| AF425643.1 | - | 6 | 760 | |||
| BC096601.1 | - | 6 | 866 | 4974190 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab40b| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139147.3 | NP_631886.2 | 6 | 872 | ||
| GenBank mRNA | AB232638.1 | - | 6 | 760 | ||
| AF425643.1 | - | 6 | 760 | |||
| BC096601.1 | - | 6 | 866 | 4974190 | ||