siRNA Details
| Gene Symbol: Defb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139225.1 | NP_631971.1 | 2 | 196 | ||
| GenBank mRNA | AJ437645.1 | - | 2 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb10| Gene Symbol: Defb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139225.1 | NP_631971.1 | 2 | 196 | ||
| GenBank mRNA | AJ437645.1 | - | 2 | 196 | ||