siRNA Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291039.1 | NP_001277968.1 | 13 | 3184 | ||
| NM_138651.7 | NP_619592.1 | 13 | 3507 | |||
| GenBank mRNA | AK036328.1 | - | 13 | 3358 | ||
| AK147541.1 | - | 13 | 3399 | |||
| AK166001.1 | - | 13 | 3358 | |||

Cds2
Genomic Map