siRNA Details
| Gene Symbol: Ufsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138668.2 | NP_619609.1 | 12 | 1490 | ||
| GenBank mRNA | AK075795.1 | - | 12 | 1485 | ||
| AK078309.1 | - | 12 | 1491 | |||
| AK160164.1 | - | 11 | 1565 | |||
| BC005503.1 | - | 12 | 1491 | 3490744 | ||

Ufsp2
Genomic Map