siRNA Details
| Gene Symbol: Mpst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162492.1 | NP_001155964.1 | 3 | 1240 | ||
| NM_001162493.1 | NP_001155965.1 | 3 | 1210 | |||
| NM_138670.3 | NP_619611.3 | 3 | 1344 | |||
| GenBank mRNA | AK136571.1 | - | 2 | 1239 | ||
| BC004079.1 | - | 2 | 1185 | 3592659 | ||
| BC006066.1 | - | 2 | 1185 | |||
| BC094469.1 | - | 3 | 1344 | 5009481 | ||

Mpst
Genomic Map