siRNA Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170848.1 | NP_001164319.1 | 9 | 1980 | ||
| NM_138680.2 | NP_619621.2 | 10 | 2010 | |||
| GenBank mRNA | AF318301.1 | - | 10 | 1935 | ||
| AK134191.1 | - | 9 | 1894 | |||
| AK147889.1 | - | 12 | 2150 | |||
| AK153527.1 | - | 13 | 2238 | |||
| BC020058.1 | - | 11 | 1735 | 4009362 | ||
| BC056354.1 | - | 10 | 2008 | 6811460 | ||
| BC056970.1 | - | 10 | 2008 | 6811460 | ||

Luc7l2
Genomic Map