siRNA Details
| Gene Symbol: Lrrc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138682.2 | NP_619623.2 | 1 | 2079 | ||
| GenBank mRNA | AF300458.1 | - | 1 | 2036 | ||
| AK046781.1 | - | 1 | 1286 | |||
| BC012209.1 | - | 1 | 2080 | |||
| BC117833.1 | - | 1 | 2020 | 40092947 | ||
| BC117834.1 | - | 1 | 2020 | 40092948 | ||
| DQ177325.1 | - | 1 | 2079 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001037336, DQ119102.

Lrrc4
Genomic Map