siRNA Details
| Gene Symbol: Cys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004455.2 | NP_001004455.2 | 5 | 1697 | ||
| NM_001161807.1 | NP_001155279.1 | 5 | 1737 | |||
| NM_138686.3 | NP_619627.3 | 5 | 1768 | |||
| GenBank mRNA | AF390548.1 | - | 5 | 1786 | ||
| AK013490.1 | - | 5 | 929 | |||
| CF613278.1 | - | 1 | 416 | |||

Cys1
Genomic Map