siRNA Details
| Gene Symbol: Asun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138757.2 | NP_620096.2 | 12 | 1831 | ||
| GenBank mRNA | AK196736.1 | - | 1 | 46 | ||
| BC026851.1 | - | 12 | 1760 | 4459471 | ||
| BC027229.1 | - | 12 | 1416 | 2615769 | ||
| CK618181.1 | - | 7 | 524 | |||

Asun
Genomic Map