siRNA Details
| Gene Symbol: Pou4f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138945.2 | NP_620395.2 | 2 | 1010 | ||
| GenBank mRNA | AK039639.1 | - | 884 | |||
| BC145941.1 | - | 2 | 1010 | 40131244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou4f3| Gene Symbol: Pou4f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138945.2 | NP_620395.2 | 2 | 1010 | ||
| GenBank mRNA | AK039639.1 | - | 884 | |||
| BC145941.1 | - | 2 | 1010 | 40131244 | ||