siRNA Details
| Gene Symbol: Rassf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138956.3 | NP_620406.1 | 5 | 3141 | ||
| GenBank mRNA | AK155495.1 | - | 5 | 3168 | ||
| AK167627.1 | - | 5 | 2973 | |||
| AK194793.1 | - | 1 | 348 | |||
| BC011511.1 | - | 5 | 3144 | 3495833 | ||
| BC071204.1 | - | 5 | 3152 | 2648929 | ||

Rassf3
Genomic Map