siRNA Details
| Gene Symbol: SEPHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012248.3 | NP_036380.2 | 1 | 1931 | ||
| GenBank mRNA | AK092003.1 | - | 3 | 1683 | ||
| AK093889.1 | - | 2 | 1688 | |||
| BC002381.2 | - | 1 | 1815 | 2961275 | ||
| BC016643.1 | - | 1 | 1804 | 3923979 | ||
| BG565156.1 | - | 1 | 311 | |||
| U43286.1 | - | 1 | 1821 | |||

SEPHS2
Genomic Map