siRNA Details
Gene Symbol: Rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139147.3 | NP_631886.2 | 6 | 1154 | ||
GenBank mRNA | AF425643.1 | - | 6 | 1041 | ||
BC096601.1 | - | 6 | 1148 | 4974190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139147.3 | NP_631886.2 | 6 | 1154 | ||
GenBank mRNA | AF425643.1 | - | 6 | 1041 | ||
BC096601.1 | - | 6 | 1148 | 4974190 |