siRNA Details
| Gene Symbol: Zfp52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144515.2 | NP_653098.2 | 5 | 669 | ||
| GenBank mRNA | AK032299.1 | - | 5 | 670 | ||
| BC021527.1 | - | 5 | 581 | 5353641 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp52| Gene Symbol: Zfp52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144515.2 | NP_653098.2 | 5 | 669 | ||
| GenBank mRNA | AK032299.1 | - | 5 | 670 | ||
| BC021527.1 | - | 5 | 581 | 5353641 | ||