siRNA Details
Gene Symbol: Angel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144524.2 | NP_653107.2 | 2 | 195 | ||
GenBank mRNA | AK143549.1 | - | 2 | 196 | ||
BC019148.1 | - | 2 | 210 | 5052974 | ||
BY194341.1 | - | 2 | 197 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
