siRNA Details
| Gene Symbol: Kazn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109684.1 | NP_001103154.1 | 5 | 1160 | ||
| NM_001109685.1 | NP_001103155.1 | 6 | 1178 | |||
| NM_144531.3 | NP_653114.3 | 5 | 1160 | |||
| GenBank mRNA | AK018497.1 | - | 6 | 1158 | ||
| AK032460.1 | - | 3 | 399 | |||
| AK052809.1 | - | 6 | 1179 | |||
| AK173090.1 | - | 5 | 1160 | |||
| BC022941.1 | - | 5 | 1164 | |||
| BC100387.1 | - | 3 | 385 | |||
| CD349067.1 | - | 5 | 735 | |||

Kazn
Genomic Map