siRNA Details
| Gene Symbol: Nmnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144533.2 | NP_653116.1 | 2, 3 | 552 | ||
| GenBank mRNA | AK041526.1 | - | 2, 3 | 281 | ||
| BC005737.1 | - | 2, 3 | 552 | 3599134 | ||
| BC092086.1 | - | 2, 3 | 261 | 5040019 | ||

Nmnat3
Genomic Map