siRNA Details
| Gene Symbol: Amhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144547.2 | NP_653130.2 | 2 | 180 | ||
| GenBank mRNA | AF503863.1 | - | 2 | 127 | ||
| BB868995.1 | - | 2 | 182 | |||
| BC119571.1 | - | 2 | 111 | 40057985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amhr2| Gene Symbol: Amhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144547.2 | NP_653130.2 | 2 | 180 | ||
| GenBank mRNA | AF503863.1 | - | 2 | 127 | ||
| BB868995.1 | - | 2 | 182 | |||
| BC119571.1 | - | 2 | 111 | 40057985 | ||