siRNA Details
| Gene Symbol: Hnrnpll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144802.4 | NP_659051.3 | 10 | 1830 | ||
| GenBank mRNA | AK012866.1 | - | 10 | 1546 | ||
| AK146879.1 | - | 10 | 1792 | |||
| BC004763.1 | - | 7 | 803 | |||
| BC006805.1 | - | 1 | 1502 | |||
| BC012849.1 | - | 10 | 1776 | 3587716 | ||
| BC038912.1 | - | 2 | 322 | |||

Hnrnpll
Genomic Map