siRNA Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139291.3 | NP_647452.1 | 4 | 400 | ||
| GenBank mRNA | AK012627.1 | - | 4 | 401 | ||
| AK047055.1 | - | 420 | ||||
| AK086692.1 | - | 879 | ||||
| AK135556.1 | - | 400 | ||||
| BC005775.1 | - | 3 | 327 | 3602527 | ||
| BC114350.2 | - | 2 | 187 | 40053418 | ||

Cdc26
Genomic Map