siRNA Details
| Gene Symbol: Otoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139310.1 | NP_647471.1 | 4, 5 | 220 | ||
| GenBank mRNA | AK157904.1 | - | 4, 5 | 252 | ||
| AY055122.1 | - | 4, 5 | 220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otoa| Gene Symbol: Otoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139310.1 | NP_647471.1 | 4, 5 | 220 | ||
| GenBank mRNA | AK157904.1 | - | 4, 5 | 252 | ||
| AY055122.1 | - | 4, 5 | 220 | |||