siRNA Details
| Gene Symbol: 2900011O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144518.3 | NP_653101.1 | 3 | 414 | ||
| GenBank mRNA | AK019307.1 | - | 3 | 407 | ||
| AK035868.1 | - | 3 | 391 | |||
| AK049997.1 | - | 3 | 446 | |||
| AK076136.1 | - | 3 | 433 | |||
| AK136859.1 | - | 3 | 414 | |||
| AK139470.1 | - | 3 | 426 | |||
| BC022741.1 | - | 3 | 393 | 4511454 | ||

2900011O08Rik
Genomic Map