siRNA Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144528.3 | NP_653111.1 | 2, 3 | 267 | ||
| GenBank mRNA | AK042319.1 | - | 2, 3 | 134 | ||
| AK154781.1 | - | 2, 3 | 233 | |||
| AK184279.1 | - | 1, 2 | 69 | |||
| BB870939.1 | - | 2, 3 | 236 | |||
| BC016543.1 | - | 2, 3 | 239 | 3158083 | ||
| BU147682.1 | - | 2, 3 | 331 | |||
| DV648665.1 | - | 2, 3 | 162 | |||

Rnf126
Genomic Map