siRNA Details
| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144543.2 | NP_653126.1 | 5 | 471 | ||
| GenBank mRNA | AB080370.1 | - | 4 | 395 | ||
| AK021335.1 | - | 5 | 425 | |||
| AK077639.1 | - | 5 | 441 | |||
| BC016594.1 | - | 5 | 425 | 2631627 | ||

Thyn1
Genomic Map