siRNA Details
| Gene Symbol: 2210407C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144544.2 | NP_653127.2 | 5 | 557 | ||
| GenBank mRNA | AA124355.1 | - | 5 | 543 | ||
| AA434919.1 | - | 3 | 363 | |||
| AY148487.1 | - | 4 | 463 | |||
| BC019553.1 | - | 5 | 535 | 4456658 | ||

2210407C18Rik
Genomic Map