siRNA Details
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290541.1 | NP_001277470.1 | 5 | 3837 | ||
| NM_139306.3 | NP_647467.1 | 6 | 3975 | |||
| GenBank mRNA | AF282864.1 | - | 6 | 3932 | ||
| AK035314.1 | - | 1 | 2870 | |||
| AK215771.1 | - | 1 | 40 | |||
| BC023423.1 | - | 1 | 1104 | |||
| BC059819.1 | - | 5 | 3837 | 6839525 | ||

Acer2
Genomic Map