siRNA Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144491.2 | NP_652762.2 | 11 | 1136 | ||
| GenBank mRNA | AK128996.1 | - | 1127 | |||
| AY078170.1 | - | 11 | 1127 | |||
| BC022738.1 | - | 6 | 555 | |||
| BC044663.1 | - | 11 | 1107 | |||
| BC080761.1 | - | 11 | 1112 | |||

Dph1
Genomic Map