siRNA Details
| Gene Symbol: Meg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018105.1 | - | 5 | 2935 | ||
| AK147321.1 | - | 7 | 4402 | |||
| BC028971.1 | - | 1 | 1215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Meg3| Gene Symbol: Meg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018105.1 | - | 5 | 2935 | ||
| AK147321.1 | - | 7 | 4402 | |||
| BC028971.1 | - | 1 | 1215 | |||