siRNA Details
| Gene Symbol: Fam64a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144526.3 | NP_653109.2 | 6 | 902 | ||
| GenBank mRNA | AK011345.1 | - | 6 | 900 | ||
| AK049710.1 | - | 6 | 903 | |||
| AK078429.1 | - | 6 | 903 | |||
| BC020115.1 | - | 6 | 888 | 3988382 | ||
| BC029115.1 | - | 6 | 888 | 3983148 | ||

Fam64a
Genomic Map