siRNA Details
| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144543.2 | NP_653126.1 | 8 | 823 | ||
| GenBank mRNA | AB080370.1 | - | 7 | 747 | ||
| AK077639.1 | - | 7 | 642 | |||
| BC016594.1 | - | 8 | 777 | 2631627 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thyn1| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144543.2 | NP_653126.1 | 8 | 823 | ||
| GenBank mRNA | AB080370.1 | - | 7 | 747 | ||
| AK077639.1 | - | 7 | 642 | |||
| BC016594.1 | - | 8 | 777 | 2631627 | ||