siRNA Details
| Gene Symbol: Zfp119a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144546.6 | NP_653129.1 | 4 | 1879 | ||
| GenBank mRNA | AF242376.1 | - | 4 | 1880 | ||
| AK047498.1 | - | 919 | ||||
| BC118950.2 | - | 4 | 1856 | 40092426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp119a| Gene Symbol: Zfp119a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144546.6 | NP_653129.1 | 4 | 1879 | ||
| GenBank mRNA | AF242376.1 | - | 4 | 1880 | ||
| AK047498.1 | - | 919 | ||||
| BC118950.2 | - | 4 | 1856 | 40092426 | ||