siRNA Details
| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172095.1 | NP_001165566.1 | 10 | 1336 | ||
| NM_144787.2 | NP_659036.1 | 10 | 1400 | |||
| GenBank mRNA | AK042792.1 | - | 9 | 1115 | ||
| AK053104.1 | - | 10 | 1276 | |||
| AK081769.1 | - | 10 | 1343 | |||
| AK144103.1 | - | 10 | 1363 | |||
| AK144890.1 | - | 10 | 1374 | |||
| AK173023.1 | - | 10 | 1400 | |||
| BC020180.1 | - | 10 | 1342 | 4923943 | ||
| BC042424.1 | - | 10 | 1336 | 5357211 | ||

Kdm4c
Genomic Map