siRNA Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144801.2 | NP_659050.2 | 7 | 1095 | ||
| GenBank mRNA | AK035836.1 | - | 6 | 952 | ||
| AK039173.1 | - | 6 | 1165 | |||
| AK087636.1 | - | 6 | 1300 | |||
| BC017618.1 | - | 7 | 1089 | 4923610 | ||
| Gene Symbol: 2900057H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013719.1 | - | 734 | |||

Tmem143
Genomic Map